ContactUsLinkComponent
J.T.Baker® BAKERBOND spe™, Frits for Filtration Columns
J.T.Baker® BAKERBOND spe™, Frits for Filtration Columns
Catalog # 7121-05
Supplier:  Avantor
J.T.Baker® BAKERBOND spe™, Frits for Filtration Columns
Catalog # 7121-05
Supplier:  Avantor
Specifications
  • Description:
    Frits for 3 ml polypropylene columns
  • Seq:
    10
  • Pk:
    50