Order Entry
Belgium
ContactUsLinkComponent
Sodium carbonate monohydrate 99.5-100.5% (by anhydrous basis), crystals NF, Multi-Compendial, J.T.Baker®
Irritant
Sodium carbonate monohydrate 99.5-100.5% (by anhydrous basis), crystals NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Sodium carbonate monohydrate 99.5-100.5% (by anhydrous basis), crystals NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    Passes Test
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Water (H₂O):
    12.0 - 15.0 %
  • NF - Assay (Na₂CO₃) (anhydrous basis):
    99.5 - 100.5 %
  • NF - Endotoxin Concentration (EU/g):
    ≤ 2.5
  • EP/BP - Assay (Na₂CO₃):
    83.0 - 87.5 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Alkali Hydroxides and Bicarbonates:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤ 125 ppm
  • EP/BP - Sulfate (SO₄):
    ≤ 250 ppm
  • EP/BP - Iron (Fe):
    ≤ 50 ppm
  • No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
    = 50 ppm

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together