

Titriplex® III ACS, ISO, Reag. Ph. Eur. analytical reagent, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:6381-92-6
:
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037
:372.24 g/mol
Merck Index:13,03543
Melting Pt:255 °C
:M-Clarity™ quality level = MQ100
EINECS:205-358-3
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:100 g/l (20°C)
- Melting Point:110 °C Elimination of water of crystallisation
- Molar Mass:700 kg/m3
- Bulk Density:700 kg/m3
- pH:4 - 5 (50 g/l, H2O, 20 °C)
- Grade:ACS,ISO,Reag. Ph Eur
- Synonyms:Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetraacetic acid disodium salt
- Hill Formula:C10H14N2Na2O8* 2 H2O
- Chemical Formula:C10H14N2Na2O8* 2 H2O
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.08418.0250_TDS_EN ProductPage_108418
Product specification
Merck Brighter titration Titripur®
Product Literature
msds_108418
SDS
Frequently Bought Together