Order Entry
Belgium
ContactUsLinkComponent
tri-Sodium citrate dihydrate 99.0-100.5% (dry basis), crystallised BP, Ph. Eur., USP
tri-Sodium citrate dihydrate 99.0-100.5% (dry basis), crystallised BP, Ph. Eur., USP
Supplier:  VWR Chemicals
Certificates
undefined
tri-Sodium citrate dihydrate 99.0-100.5% (dry basis), crystallised BP, Ph. Eur., USP
Supplier:  VWR Chemicals
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Conforms (see CoA/CoS)
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Residual solvents:
    Passes test
  • As (Arsenic):
    Max. 1 ppm
  • Cd (Cadmium):
    Max. 1 ppm
  • Co (Cobalt):
    Max. 1 ppm
  • Hg (Mercury):
    Max. 2 ppm
  • Ni (Nickel):
    Max. 1 ppm
  • Pb (Lead):
    Max. 3 ppm
  • V (Vanadium):
    Max. 2 ppm
  • Conforms to Ph.Eur./USP/BP:
    Passes test
  • Ph.Eur./BP specifications::
    .
  • Assay (on anhydrous) [Ph.Eur./BP]:
    99,0 --> 101,0 %
  • Identification A:
    Passes test Ph.Eur./BP
  • Identification B:
    Passes test Ph.Eur./BP
  • Appearance of solution:
    Passes test Ph.Eur./BP
  • Acidity or alkalinity:
    Passes test Ph.Eur./BP
  • Readily carbonisable substances:
    Passes test Ph.Eur./BP
  • Cl (Chloride) [Ph.Eur./BP]:
    Max. 50 ppm
  • C2O4 (Oxalate) [Ph.Eur./BP]:
    Max. 300 ppm
  • SO4 (Sulphate) [Ph.Eur./BP]:
    Max. 150 ppm
  • Water [Ph.Eur./BP]:
    11,0 --> 13,0 %
  • USP specifications::
    .
  • Assay (calculated on anhydrous) [USP]:
    99,0 --> 100,5 %
  • Identification C:
    Passes test USP
  • Tartrate:
    Passes test USP
  • Alkalinity:
    Passes test USP
  • Water determination [USP]:
    10,0 --> 13,0 %

Specification Test Results

Frequently Bought Together