Order Entry
Belgium
ContactUsLinkComponent
tri-Sodium citrate dihydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
tri-Sodium citrate dihydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
tri-Sodium citrate dihydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Limited Time Sale
Select items below are currently on sale

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    425 g/l (25 °C)
  • Melting Point:
    150 °C (anhydrous substance)
  • Molar Mass:
    294.10 g/mol
  • Bulk Density:
    900 - 1100 kg/m3
  • Density:
    1.76 g/cm3
  • pH:
    7.5 - 9.5 (50 g/l, H2O, 20 °C)

Specification Test Results

Frequently Bought Together