tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), granular USP, FCC, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate
CAS Number:6132-04-3
:
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031
Density:1.70 g/cm³
Molecular Weight:294.1 g/mol
Melting Pt:150
Merck Index:13,08675
Storage Temperature:Ambient
EINECS:200-675-3
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Meets F.C.C. Requirements:99.0 - 100.5%
- Food GMP Manufactured Product:Passes Test
- Meets U.S.P Requirements:Passes Test
- GMP Manufactured Product:Passes Test
- CAUTION: For Manufacturing, processing or repackaging:Passes Test
- Bulk Pharmaceutical Chemical:10.0 - 13.0%
- Bulk Food Chemical:Passes Test
- USP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis):99.0 - 100.5 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Identification C:Passes Test
- USP - Alkalinity:Passes Test
- USP - Water (H₂O):10.0 - 13.0 %
- USP - Tartrate:Passes Test
- FCC - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis):99.0 - 100.5 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Alkalinity:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Water (FCC):10.0 - 13.0 %
Specification Test Results
Related Information
Frequently Bought Together