Order Entry
Belgium
ContactUsLinkComponent
tri-Sodium citrate dihydrate 99.0-105.0% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®
tri-Sodium citrate dihydrate 99.0-105.0% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
tri-Sodium citrate dihydrate 99.0-105.0% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • GMP Manufactured Product:
    Passes Test
  • Meets E.P. Chemical Specifications:
    Passes Test
  • Meets B.P. Chemical Specifications:
    Passes Test
  • Meets U.S.P Requirements:
    Passes Test
  • Meets J.P. Chemical Specifications:
    10.0 - 13.0%
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    99.0 - 100.5%
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Identification C:
    Passes Test
  • USP - Alkalinity:
    Passes Test
  • USP - Water (H₂O):
    10.0 - 13.0 %
  • USP - Tartrate:
    Passes Test
  • USP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis):
    99.0 - 100.5 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Acidity or Alkalinity:
    Passes Test
  • EP/BP - Readily Carbonizable Substances:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤50 ppm
  • EP/BP - Oxalate (C₂O₄):
    ≤300 ppm
  • EP/BP - Sulfate (SO₄):
    ≤150 ppm
  • EP/BP - Water (H₂O):
    11.0 - 13.0 %
  • EP/BP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis):
    99.0 - 101.0 %
  • EP/BP - Endotoxin Concentration (EU/g):
    ≤2.5
  • JP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis):
    99.0 - 101.0 %
  • JP - pH (1 in 20):
    7.5 - 8.5
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Chloride (Cl):
    ≤0.015 %
  • JP - Identification:
    Passes Test
  • JP - Sulfate (SO₄):
    ≤0.048 %
  • JP - Heavy Metals (as Pb):
    ≤10 ppm
  • JP - Arsenic (As):
    ≤2 ppm
  • JP - Tartrate:
    Passes Test
  • JP - Oxalate (C₂O₄):
    Passes Test
  • JP - Readily Carbonizable Substances:
    Passes Test
  • JP - Loss on Drying at 180°C:
    10.0 - 13.0 %
  • E. coli:
    Negative
  • P. aureginosa:
    Negative

Specification Test Results

Related Information

Frequently Bought Together