tri-Sodium citrate dihydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
: Avantor
: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate
:6132-04-3
:
:Na₃C₆H₅O₇·2H₂O
:MFCD00150031
:1.76 g/cm³ (20 °C)
:294.1 g/mol
:150
:13,08675
:Ambient
:200-675-3
- Environmentally Preferable:
- Assay:min. 99.0 %
- Calcium (Ca):max. 0.005 %
- Chloride (Cl):max. 0.001 %
- Insoluble Matter:max. 0.005 %
- pH of 5% Solution at 25°C:7.0 - 9.0
- Sulfate (SO4):max. 0.002 %
- Trace Impurities (in ppm) Iron (Fe):max. 5
- Trace Impurities (in ppm) Heavy Metals (as Pb):max. 5
Frequently Bought Together