Order Entry
Belgium
ContactUsLinkComponent
 
 
Citric acid monohydrate 99.5-100.5% (by anhydrous basis), granular, GenAR® USP, Macron Fine Chemicals™
 
undefined
 
Citric acid monohydrate 99.5-100.5% (by anhydrous basis), granular, GenAR® USP, Macron Fine Chemicals™
 

 

  • USP - Assay (as C₆H₈O₇) (calculated on anhydrous basis):
    99.5 - 100.5 %
  • USP - Clarity of Solution:
    Passes Test
  • USP - Color of Solution:
    Passes Test
  • USP - Identification:
    Passes Test
  • USP - Water (H₂O):
    7.5 - 9.0 %
  • USP - Residue on Ignition:
    ≤0.1 %
  • USP - Readily Carbonizable Substances:
    Passes Test
  • USP - Oxalic Acid:
    ≤0.036 %
  • USP - Sulfate (SO₄):
    ≤0.015 %
  • EP/BP - Assay (as C₆H₈O₇) (calculated on anhydrous basis):
    99.5 - 100.5 %
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification E:
    Passes Test
  • EP/BP - Oxalic Acid:
    ≤360 ppm
  • EP/BP - Sulfate (SO₄):
    ≤150 ppm
  • EP/BP - Ash (sulfated):
    ≤0.1 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Readily Carbonizable Substances:
    Passes Test
  • EP/BP - Water (H₂O):
    7.5 - 9.0 %
  • JP - Assay (C₆H₈O₇) (anhydrous basis):
    99.5 - 100.5 %
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤10 ppm
  • JP - Identification:
    Passes Test
  • JP - Oxalic Acid:
    ≤360 ppm
  • JP - Readily Carbonizable Substances:
    Passes Test
  • JP - Residue on Ignition:
    ≤0.10 %
  • JP - Sulfate (SO₄):
    ≤150 ppm
  • JP - Water (H₂O):
    7.5 - 9.0 %
  • Endotoxin Concentration (IU/mg):
    ≤0.5

 

Frequently Bought Together