
Citric acid monohydrate 99.5-100.5% (by anhydrous basis), granular USP, Macron Fine Chemicals™
:5949-29-1
:
:C₆H₈O₇.H₂O
:MFCD00149972
:210.14 g/mol
:13,02350
:135…152 °C
:1.552 g/cm³ (20 °C)
:173.9 °C (closed cup)
:201-069-1
:GHS07
:Ambient
- Meets U.S.P Requirements:—
- GMP Manufactured Product:—
- Preserve in Tight Containers:—
- CAUTION: For Manufacturing, processing or repackaging:—
- Bulk Pharmaceutical Chemical:—
- USP - Assay (as C₆H₈O₇) (calculated on anhydrous basis):99.5 - 100.5 %
- USP - Clarity of Solution:Passes Test
- USP - Color of Solution:Passes Test
- USP - Identification:Passes Test
- USP - Water (H₂O)(by Karl Fischer titrn):7.5 - 9.0 %
- USP - Residue on Ignition:≤0.1 %
- USP - Readily Carbonizable Substances:Passes Test
- USP - Sulfate (SO₄):≤0.015 %
- USP - Limit of Oxalic Acid:≤0.036 %
- Solubility (5 in 50):Passes Test
Frequently Bought Together