di-Ammonium oxalate monohydrate ≥99%, GPR RECTAPUR®
Catalog # 21288.266
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 99 %
- Heavy metals (as Pb):Max. 20 ppm
- Ignition residue (SO4):Max. 0.1 %
- Fe (Iron):Max. 20 ppm
Specification Test Results
About this item
Synonyms: Ammonium oxalate monohydrate, Ammonium oxalate-1-hydrate, Oxalic acid diammonium salt monohydrate, Oxalic acid ammonium salt monohydrate, Ammonium ethanedioate monohydrate, Ammonium Oxalate, Monohydrate
:
:142.11 g/mol
Merck Index:13,00541
Boiling Pt:70 °C (1013 hPa)
Melting Pt:70 °C
Density:∼1.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:214-202-3
UN:3288
ADR:6.1,III
:GHS07
CAS Number:6009-70-7
Formula:(NH₄)₂C₂O₄·H₂O
MDL Number:MFCD00149694