Order Entry
Belgium
ContactUsLinkComponent
Sodium acetate trihydrate 99.0-101.0%, crystals, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Sodium acetate trihydrate 99.0-101.0%, crystals, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium acetate trihydrate 99.0-101.0%, crystals, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    99.0 - 101.0 %
  • Assay (CH₃COONa·3H₂O):
    99.0 - 101.0 %
  • DNase Activity (None Detected):
    Passes Test
  • RNase Activity:
    Passes Test
  • Protease Activity:
    Passes Test
  • pH of 5% Solution at 25°C:
    7.5 - 9.2
  • Absorbance of a 1M Solution - 254 nm:
    ≤0.02
  • Absorbance of a 1M Solution - 280 nm:
    ≤0.01
  • Absorbance of a 1M Solution - 350 nm:
    ≤0.01
  • Insoluble Matter:
    ≤0.005 %
  • Chloride (Cl):
    ≤0.001 %
  • Substances Reducing Permanganate:
    Passes Test
  • Sulfate (SO₄):
    ≤0.002 %
  • Calcium, Magnesium, and R₂O₃ Precipitate:
    ≤0.01 %
  • Potassium (K):
    ≤0.005 %
  • Phosphate (PO₄):
    ≤5 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together