Order Entry
Belgium
ContactUsLinkComponent
Trifluoroacetic acid 0.05% (v/v) in acetonitrile, LiChrosolv® for HPLC, Supelco®
Trifluoroacetic acid 0.05% (v/v) in acetonitrile, LiChrosolv® for HPLC, Supelco®
Catalog # 4.80672.2500
Supplier:  Merck
CAS Number:  
undefined
Trifluoroacetic acid 0.05% (v/v) in acetonitrile, LiChrosolv® for HPLC, Supelco®
Catalog # 4.80672.2500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :114.02 g/mol
ADR:3,II
 :M-Clarity™ quality level = MQ100
EINECS:200-929-3
UN:1648
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169