Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-2-Aminoethyl-1-naphthylamine dihydrochloride
N-(1-Naphthyl)ethylenediamine dihydrochloride
N-(1-Naphthyl)ethylenediamine dihydrochloride
SERA30060.01
:
Serva
:
N-(1-Naphthyl)ethylenediamine dihydrochloride
SERA30060.01
:
Serva
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
259.18 g/mol
:
12,06496
:
188…190 °C
:
215-981-2
:
GHS07
:
Ambient
:
1465-25-4
:
C₁₂H₁₆Cl₂N₂
:
MFCD00012556