
N-(1-Naphthyl)ethylenediamine dihydrochloride
Catalog # MOLEM69625605
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:259.18 g/mol
Merck Index:12,06496
Melting Pt:188…190 °C
EINECS:215-981-2
:GHS07
Storage Temperature:Ambient
CAS Number:1465-25-4
Formula:C₁₂H₁₆Cl₂N₂
MDL Number:MFCD00012556