Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Gentamicin sulphate
Gentamicin sulphate 50 mg/ml, Reagent Grade
Gentamicin sulphate 50 mg/ml, Reagent Grade
Gentamicin sulphate 50 mg/ml, Reagent Grade
Catalog # ICNA0916762J8
Supplier:
MP Biomedicals
CAS Number:
Expand image
Gentamicin sulphate 50 mg/ml, Reagent Grade
Catalog # ICNA0916762J8
Supplier:
MP Biomedicals
CAS Number:
Specifications
Pk:
100 mL
Seq:
2440
Specifications
About this item
Product Details
:
CAS Number:
1405-41-0
MDL Number:
MFCD00270181
Product Details