
Guanidinium hydrochloride, Millipore®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
CAS Number:50-01-1
:
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026
:95.53 g/mol
Merck Index:13,04578
Melting Pt:185 °C
Density:1.354 g/cm³ (20 °C)
EINECS:200-002-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together