Order Entry
Belgium
ContactUsLinkComponent
Guanidinium hydrochloride ≥99.5% for biotechnology
Irritant
Guanidinium hydrochloride ≥99.5% for biotechnology
Catalog # 0287-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Guanidinium hydrochloride ≥99.5% for biotechnology
Catalog # 0287-500G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bottle, amber
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Solubility (6 M, Water):
    Pass
  • Absorbance 260 nm (6 M, Water):
    ≤0.03
  • Arsenic:
    ≤0.0005%
  • DNase:
    None Detected
  • Iron:
    ≤0.0005%
  • Lead:
    <0.0005%
  • Melting Point:
    181 - 187 °C
  • Protease:
    None Detected
  • Purity:
    >99.5%
  • RNase:
    None

Specification Test Results

About this item

 :
 :95.53 g/mol
Merck Index:13,04578
Melting Pt:185 °C
Density:1.354 g/cm³ (20 °C)
EINECS:200-002-3
 :GHS07
Storage Temperature:Ambient
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026