Order Entry
Belgium
Orders LinkContactUsLinkComponent
Basic fuchsin, Sigma-Aldrich®

Basic fuchsin, Sigma-Aldrich®

Health
Basic fuchsin, Sigma-Aldrich®
Certificates
Basic fuchsin, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
  • CAS Number:632-99-5
  • Formula:C₂₀H₂₀ClN₃
  • MDL Number:MFCD00012569
  • Molecular Weight:337.85 g/mol
  • Merck Index:12,05686
  • Melting Pt:235 °C
  • Density:1.22 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:211-189-6

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 148,00 €
    148,00 €
    1 * 5 g
       148,00 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 577,00 €
    577,00 €
    1 * 25 g
       577,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.