Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Basic fuchsin

Glass bottle for solids, 10 g
Catalog # OXOIBR0050A
Health
Basic fuchsin
Catalog # OXOIBR0050A
CAS Number:  632-99-5
Basic fuchsin

Glass bottle for solids, 10 g

CAS Number:  632-99-5
Price
38,70 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
  • CAS Number:632-99-5
  • Formula:C₂₀H₂₀ClN₃
  • MDL Number:MFCD00012569
  • Molecular Weight:337.85 g/mol
  • Merck Index:12,05686
  • Melting Pt:235 °C
  • Density:1.22 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:211-189-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications