Basic fuchsin
Glass bottle for solids, 25 g
Catalog # MOLE51119322-25G
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
- CAS Number:632-99-5
- Formula:C₂₀H₂₀ClN₃
- MDL Number:MFCD00012569
- Molecular Weight:337.85 g/mol
- Merck Index:12,05686
- Melting Pt:235 °C
- Density:1.22 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:211-189-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



