Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Basic fuchsin

Health
Irritant
Basic fuchsin
Certificates
Basic fuchsin

3 Options Available Below

Product Details & Documents

CAS No.: 632-99-5 Molecular Formula: C20H20ClN3 Formula Weight: 337.86 Melting Point: 268-270° dec. Solubility: Soluble in water Beilstein Reference: 4166684 Merck Reference: 14,5652 MDL No.: MFCD00012569

Synonyms:  CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
  • CAS Number:632-99-5
  • Formula:C₂₀H₂₀ClN₃
  • MDL Number:MFCD00012569
  • Molecular Weight:337.85 g/mol
  • Merck Index:14,5652
  • Melting Pt:268…270 °C (dec.)
  • Density:1.22 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:211-189-6
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.