Order Entry
Belgium
Orders LinkContactUsLinkComponent
Basic fuchsin 70+%, pure, certified

Basic fuchsin 70+%, pure, certified

Irritant
Health
Basic fuchsin 70+%, pure, certified
Certificates
Basic fuchsin 70+%, pure, certified

3 Options Available Below

Product Details & Documents
Synonyms:  CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
  • CAS Number:632-99-5
  • Formula:C₂₀H₂₀ClN₃
  • MDL Number:MFCD00012569
  • Molecular Weight:337.85 g/mol
  • Merck Index:12,05686
  • Melting Pt:250 °C (dec.)
  • Density:1,22 g/cm³ (20 °C)
  • Flash Pt:200 °C
  • Storage Temperature:Ambient
  • EINECS:211-189-6

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Dark green Crystalline powder
  • Infrared spectrum:
    Conforms
  • Dye content:
    ≥70.0 %
  • UV Lambda max:
    547 to 552 nm (in 50% ethanol)
  • Absorption Ratio P-15/P+15:
    1.16 to 1.35
  • BSC Certification:
    Passes
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 49,70 €
    49,70 €
    1 * 25 g
       49,70 €

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 145,00 €
    145,00 €
    1 * 100 g
       145,00 €

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 485,00 €
    485,00 €
    1 * 500 g
       485,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.