
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
Catalog # MOLEM76530445
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:237.17 g/mol
Merck Index:12,09367
Melting Pt:227…230 °C
EINECS:211-274-8
:GHS07
Storage Temperature:Ambient
CAS Number:637-01-4
Formula:C₁₀H₁₈Cl₂N₂
MDL Number:MFCD00012482