
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:637-01-4
:
Formula:C₁₀H₁₈Cl₂N₂
MDL Number:MFCD00012482
:237.17 g/mol
Merck Index:12,09367
Melting Pt:227…230 °C
:M-Clarity™ quality level = MQ200
EINECS:211-274-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:227 - 230 °C (decomposition)
- Molar Mass:237.17 g/mol
Specification Test Results
Related Information
Frequently Bought Together