Order Entry
Belgium
ContactUsLinkComponent
N,N-Dimethyl-1,4-phenylenediammonium dichloride ≥99.0% analytical reagent
Toxic
N,N-Dimethyl-1,4-phenylenediammonium dichloride ≥99.0% analytical reagent
Catalog # 23484.132
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
N,N-Dimethyl-1,4-phenylenediammonium dichloride ≥99.0% analytical reagent
Catalog # 23484.132
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Ignition residue (SO4):
    Max. 0.1 %

Specification Test Results

About this item

 :
 :209.12 g/mol
Merck Index:13,03280
Melting Pt:208…212 °C
Storage Temperature:Ambient
EINECS:208-635-7
UN:2811
ADR:6.1,II
 :GHS06
CAS Number:536-46-9
 :MFCD00012990
Formula:(CH₃)₂NC₆H₄NH₂·2HCl
MDL Number:MFCD00012991