Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Spermidine trihydrochloride
Spermidine trihydrochloride, Millipore®
Spermidine trihydrochloride, Millipore®
56766-5
:
Merck Millipore (Calbiochem)
:
Spermidine trihydrochloride, Millipore®
56766-5
:
Merck Millipore (Calbiochem)
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
254.63 g/mol
:
12,08893
:
256…258 °C
:
Freezer
:
206-379-0
:
334-50-9
:
C₇H₂₂Cl₃N₃
:
MFCD00012918