Order Entry
Belgium
ContactUsLinkComponent
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), Supelco®
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), Supelco®
Catalog # 93313-1L
Supplier:  Merck
CAS Number:  
undefined
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), Supelco®
Catalog # 93313-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :157.6 g/mol
Merck Index:12,09902
Melting Pt:150…152 °C
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:214-684-5
CAS Number:1185-53-1
Formula:NH₂C(CH₂OH)₃·HCl
MDL Number:MFCD00012590