Order Entry
Belgium
ContactUsLinkComponent
 
Corrosive
Sodium hydroxide concentrate for dilution to 1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4689.
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Sodium hydroxide concentrate for dilution to 1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4689.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    210 mL
  • Seq:
    2545

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
  • Normality (by titrimetry):
    Passes Test
  • (Makes 1N solution after dilution to 1000 mL):
  • Low in Carbonate:

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:∼105 °C (1013 hPa)
Density:1.327 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548