Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide concentrate for dilution to 1 L, 0.5 M NaOH (0.5 N) volumetric solution, Fluka™
Corrosive
Sodium hydroxide concentrate for dilution to 1 L, 0.5 M NaOH (0.5 N) volumetric solution, Fluka™
Catalog # 38217-1EA
CAS Number:  
Corrosive
Sodium hydroxide concentrate for dilution to 1 L, 0.5 M NaOH (0.5 N) volumetric solution, Fluka™
Catalog # 38217-1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:120 °C
Density:1.350 g/cm³
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548