Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution
Corrosive
Sodium hydroxide concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution
Catalog # 88623.LQ
CAS Number:  
Corrosive
Sodium hydroxide concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution
Catalog # 88623.LQ
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:1310-73-2
 :
Formula:NaOH
MDL Number:MFCD00003548
 :40 g/mol
Boiling Pt:∼105 °C (1013 hPa)
Density:1.327 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05