Order Entry
Belgium
ContactUsLinkComponent
Potassium hydroxide 1 mol/l in aqueous solution, Titrisol®, Supelco®
Corrosive
Irritant
Potassium hydroxide 1 mol/l in aqueous solution, Titrisol®, Supelco®
Catalog # 1.09918.0001
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
Potassium hydroxide 1 mol/l in aqueous solution, Titrisol®, Supelco®
Catalog # 1.09918.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Fibre case
  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.58 g/cm3 (20 °C)
  • pH:
    13.5 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

 :
 :56.11 g/mol
ADR:8,
 :M-Clarity™ quality level = MQ100
EINECS:215-181-3
UN:1814
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553