
Sulphuric acid 0.5 mol concentrated aqueous solution 0.5 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09981.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Pk:1 Ampoul
- Seq:0501
Specifications
Specification Test Results
- Solubility:(20 °C) soluble,(caution ! development of heat)
- Boiling Point:135 °C
- Density:1.29 g/cm3 (20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
About this item
:
:98.08 g/mol
ADR:8,
:M-Clarity™ quality level = MQ100
EINECS:231-639-5
UN:2796
:GHS05
Storage Temperature:Ambient
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589