Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid concentrate for dilution to 1L, 0.25 M H2SO4 (0.5N) volumetric solution, Fluka™
Corrosive
Sulphuric acid concentrate for dilution to 1L, 0.25 M H2SO4 (0.5N) volumetric solution, Fluka™
Catalog # 38295-1EA
CAS Number:  
Corrosive
Sulphuric acid concentrate for dilution to 1L, 0.25 M H2SO4 (0.5N) volumetric solution, Fluka™
Catalog # 38295-1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :98.08 g/mol
Boiling Pt:>100 °C (1.013 hPa)
Density:1.180 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589