
Hydrochloric acid concentrate for dilution to 0.1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4655.
Supplier: Avantor
CAS Number:
Specifications
- Pk:1 Ampoul
- Seq:0501
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Normality (by titrimetry):Passes Test
Specification Test Results
About this item
:
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,II
:36.46 g/mol
UN:1789
:GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324