L(+)-Histidine monochlorhydrate monohydraté, Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:5934-29-2
:
Formula:C₆H₉N₃O₂·HCl·H₂O
MDL Number:MFCD00151027
:209.63 g/mol
Merck Index:13,04741
Melting Pt:259 °C
Density:1,485 g/cm³ (20 °C)
:M-Clarity™ (Niveau de qualité) = MQ100
Storage Temperature:Température ambiante
EINECS:211-438-9
Related Information
Frequently Bought Together