Order Entry
Belgium
ContactUsLinkComponent
L(+)-Cysteine hydrochloride monohydrate 98.5-101.0% for biochemistry
L(+)-Cysteine hydrochloride monohydrate 98.5-101.0% for biochemistry
Catalog # 23257.183
Supplier:  VWR Chemicals
CAS Number:  
undefined
L(+)-Cysteine hydrochloride monohydrate 98.5-101.0% for biochemistry
Catalog # 23257.183
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    98.5 - 101.0 %
  • Identification:
    Passes test
  • Spec.opt.rot.(80 g/l;HCl 250 g/l; dried):
    5.5 - 7.0 °
  • Heavy metals (as Pb):
    ≤ 10 ppm
  • Loss on drying (Vacuum<0.7 kPa, 24 h):
    8.0 - 12.0 %
  • SO₄ (Sulphate):
    ≤ 300 ppm
  • Ninhydrin-positive substances (LC)::
    Max. 0.02 %
  • Ammonium (570 nm):
    ≤ 0.02 %
  • Any impurity:
    ≤ 0.2 %
  • Impurity A (570 nm):
    ≤ 0.5 %
  • Total impurities:
    ≤ 1.0 %

Specification Test Results

About this item

 :
 :175.64 g/mol
Merck Index:13,02810
Melting Pt:168…170 °C
Storage Temperature:Ambient
EINECS:200-157-7
CAS Number:7048-04-6
Formula:HSCH₂CH(NH₂)COOH·HCl·H₂O
MDL Number:MFCD00065606