Order Entry
Belgium
ContactUsLinkComponent
Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)
Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)
Catalog # A4228.5000PE
Supplier:  PanReac AppliChem
CAS Number:  
Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)
Catalog # A4228.5000PE
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    5 L
  • Description:
    TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer)

Specifications

About this item