Order Entry
Belgium
ContactUsLinkComponent
Buffer, TBE (Tris-borate-EDTA) powder for 10X concentrated solution
Buffer, TBE (Tris-borate-EDTA) powder for 10X concentrated solution
Catalog # A4348.9010
Supplier:  PanReac AppliChem
CAS Number:  
Buffer, TBE (Tris-borate-EDTA) powder for 10X concentrated solution
Catalog # A4348.9010
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    10 L
  • Description:
    TBE buffer solid for 10X concentrated solution (TRIS-borate-EDTA buffer)

Specifications

About this item

Powder for 10X concentrated aqueous solution (170,28 g/L). Composed of 0,89 M Tris, 0,89 M boric acid and 0,02 M EDTA.Na₂.