Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), Flacon Verre, 1 L
Catalog # MOLEM79689066
Marketsource
Supplier: Molekula
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Catalog # MOLEM79689066
Supplier: Molekula
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Type de conditionnement:Flacon Verre
- Cdt:1 L
- Désignation:TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



