Order Entry
Belgium
ContactUsLinkComponent
VWR® TBE Buffer Solution, 10X Concentrate (Tris-Borate-EDTA Buffer), UltraPure Grade
VWR® TBE Buffer Solution, 10X Concentrate (Tris-Borate-EDTA Buffer), UltraPure Grade
Catalog # E442-500ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® TBE Buffer Solution, 10X Concentrate (Tris-Borate-EDTA Buffer), UltraPure Grade
Catalog # E442-500ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    TBE buffer solution 10X concentrate (Tris-borate-EDTA buffer)

Specifications

About this item

pH (1:10) at 25 °C: 8,20 to 8,40