Order Entry
Belgium
ContactUsLinkComponent
 
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer), MP Biomedicals
Catalog # ICNA11TAE50X01
Supplier:  MP Biomedicals
CAS Number:  
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer), MP Biomedicals
Catalog # ICNA11TAE50X01
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 L
  • Description:
    TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)

Specifications

About this item