Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)

Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)

TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 1 L
N° de catalogue MOLE12439469-1L
Marketsource
Fournisseur:   Molekula
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
N° de catalogue MOLE12439469-1L
Fournisseur:  Molekula
Marketsource
Numéro CAS:  
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)

TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 1 L

Numéro CAS:  
Prix
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)

Specifications