Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer) pH 8,5

Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer) pH 8,5

TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer) pH 8,5, Glass bottle, 1 L
Catalog # A4686.1000
Supplier:   PanReac AppliChem
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer) pH 8,5
Catalog # A4686.1000
Supplier:  PanReac AppliChem
CAS Number:  
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer) pH 8,5

TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer) pH 8,5, Glass bottle, 1 L

CAS Number:  
Price
98,20 €
Log in for availability
1 * 1 L
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer) pH 8,5

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.