Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)

Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)

TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 1 L
Catalog # MOLE22306903-1L
Marketsource
Supplier:   Molekula
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)
Catalog # MOLE22306903-1L
Supplier:  Molekula
Marketsource
CAS Number:  
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)

TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 1 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.