Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ammonia solution 28%, BAKER ANALYZED® ACS, J.T.Baker®

Ammonia solution 28%, BAKER ANALYZED® ACS, J.T.Baker®

Corrosive
Irritant
Environment
Supplier:   Avantor
Ammonia solution 28%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
Ammonia solution 28%, BAKER ANALYZED® ACS, J.T.Baker®

3 Options Available Below

Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.88…0.91 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications for Product Family

Specification Test Results

  • Assay (as NH3) (acidimetric):
    28.0-30.0 %
  • Appearance:
    passes test
  • Carbon Dioxide (CO2):
    max. 0,002 %
  • Chloride (Cl):
    max. 0,5 ppm
  • Color (APHA):
    max. 5
  • Residue after Ignition:
    max. 0,001 %
  • Specific Gravity at 60° / 60°F:
    0.896-0.902
  • Substances Reducing KMnO4:
    passes test
  • Trace Impurities (in ppm):
    -
  • Aluminium (Al):
    max. 0,2
  • Arsenic and Antimony (as As):
    max. 3
  • Barium (Ba):
    max. 0,3
  • Boron (B):
    max. 0,05
  • Chromium (Cr):
    max. 0,1
  • Copper (Cu):
    max. 0,1
  • Heavy Metals (as Pb):
    max. 0,5
  • Iron (Fe):
    max. 0,1
  • Lead (Pb):
    max. 0,2
  • Magnesium (Mg):
    max. 0,2
  • Manganese (Mn):
    max. 0,1
  • Nickel (Ni):
    max. 0,1
  • Nitrates (NO3):
    max. 2
  • Phosphate (PO4):
    max. 0,4
  • Sulphate (SO4):
    max. 2
  • Tin (Sn):
    max. 0,1
  • Titanium (Ti):
    max. 0,1
  • Zinc (Zn):
    max. 0,1
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.