Ammonia solution 25%
Marketsource
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00066650
- Molecular Weight:35.05 g/mol
- Boiling Pt:32…37.7 °C (1013 hPa)
- Melting Pt:–60…–57.5 °C
- Density:0.88…0.91 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-647-6
- UN:2672
- ADR:8,III
4 Options Available Below
Product Details & Documents
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00066650
- Molecular Weight:35.05 g/mol
- Boiling Pt:32…37.7 °C (1013 hPa)
- Melting Pt:–60…–57.5 °C
- Density:0.88…0.91 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-647-6
- UN:2672
- ADR:8,III
Certificates
Search by Lot NumberProduct Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



