Tetramethylammonium hydroxide 25% (w/w) in methanol
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-59-2
:
Formula:C₄H₁₃NO
MDL Number:MFCD00008280
:91.15 g/mol
Merck Index:14,9224
Density:0.865
Flash Pt:12 °C (54 °F)
EINECS:200-882-9
UN:2924
ADR:3,II
:GHS02
:GHS06
:GHS08
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together