Order Entry
Belgium
ContactUsLinkComponent
Tetrabutylammonium hydroxide 40% in aqueous solution
Corrosive
Tetrabutylammonium hydroxide 40% in aqueous solution
Catalog # MOLE12269822-100ML
Supplier:  Molekula
CAS Number:  
Corrosive
Tetrabutylammonium hydroxide 40% in aqueous solution
Catalog # MOLE12269822-100ML
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :259.48 g/mol
Storage Temperature:Refrigerator
EINECS:218-147-6
UN:3267
ADR:8,II
 :GHS05
CAS Number:2052-49-5
Formula:C₁₆H₃₇NO
MDL Number:MFCD00009425