
Potassium iodide 5% in aqueous solution
Supplier: PROLABO CMP
Certificates
About this item
CAS Number:7681-11-0
:
Formula:Kl
MDL Number:MFCD00011405
:166 g/mol
EINECS:231-659-4
:GHS08
Storage Temperature:Ambient
Frequently Bought Together
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals